missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanes, 99%, for spectroscopy ACS, mixture of isomers, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 411555000

 View more versions of this product

Catalog No. AC411555000


 

Specifications Description & Specifications

Specifications

≥98.5% (sum of 5 isomers, total hexanes, plus methylcyclopentane)
Liquid
500mL
EcoSafPak™ Glass Bottle
14, 4694
0.01 max. at 280nm
Vapor Pressure: 160mbar at 20°C
99%
MFCD00009520
Passes Test
0.005% max.
Mixture of isomers, for spectroscopy
86.18
ACS Grade
Clear
0.6590g/mL
01, 142
92112-69-1,110-54-3
C6H14
CH3(CH2)4CH3
0.001% max.
0.0003 meq/g max.
Hex
Hexanes

Chemical Identifiers

92112-69-1,110-54-3
C6H14
86.18
Hex
≥98.5% (sum of 5 isomers, total hexanes, plus methylcyclopentane)
99%
CH3(CH2)4CH3
MFCD00009520
Hexanes, Mixture of isomers, for spectroscopy
ACS Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-777-6

RTECSNumber : MN9275000

TSCA : TSCA