missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanes, 99%, pure, mixture of isomers, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC411540100

 View more versions of this product

Catalog No. AC411540100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Hexanes
92112-69-1,110-54-3
Pure
≥98.5% (sum of isomers)
C6H14
86.18
01, 142
Hex
Authentic
1.3770 to 1.3810 (20°C, 589nm)
10L
Mixture of Isomers
Vapor Pressure: 160mbar at 20°C
Liquid
99%
CH3(CH2)4CH3
MFCD00009520
14, 4694
0.6590g/mL
Plastic Jerrycan
Clear colorless

Chemical Identifiers

92112-69-1,110-54-3
C6H14
86.18
Hex
≥98.5% (sum of isomers)
99%
CH3(CH2)4CH3
MFCD00009520
Hexanes, Mixture of Isomers
Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement
Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.

Danger