Please login to your online account to display your discounted pricing

Hexanes, 99%, pure, mixture of isomers, ACROS Organics™


Quantity 1L
Packaging EcoSafPak™ Glass Bottle

Manufacturer: thermo fisher scientific  411540010

 View more versions of this product

Catalog No. AC411540010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (sum of isomers)
Physical Form Liquid
Formula Weight 86.18
Quantity 1L
Color Clear colorless
Packaging EcoSafPak™ Glass Bottle
Purity Grade Pure
Density 0.6590g/mL
Beilstein 01, 142
Merck Index 14, 4694
Infrared Spectrum Authentic
Name Note Mixture of Isomers
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Synonym Hex
Health Hazard 1 Danger
Chemical Name or Material Hexanes
Additional Information Vapor Pressure: 160mbar at 20°C
Refractive Index 1.3770 to 1.3810 (20°C, 589nm)
Assay Percent Range 99%
MDL Number MFCD00009520
CAS 92112-69-1
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
CAS 110-54-3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hexanes
Name Note Mixture of Isomers
CAS 92112-69-1
CAS 110-54-3
Purity Grade Pure
Assay ≥98.5% (sum of isomers)
Assay Percent Range 99%
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Formula Weight 86.18
MDL Number MFCD00009520
Synonym Hex
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H

SDS & Safety Documents