Please login to your online account to display your discounted pricing

Hexanes, 99% n-hexane, ULTRA RESI-ANALYZED™, J.T.Baker™

High purity solvents and salts developed for organic residue extraction/concentration procedures

$745.63 - $2,618.17

Chemical Identifiers

Chemical Name or Material Hexanes
CAS 110-54-3
Purity Grade ULTRA RESI-ANALYZED™
Molecular Formula CH3(CH2)4CH3
Formula Weight 86.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002992

 
avantor performance
N16802
Narrow Mouth Amber Glass Bottle 1L Each for $745.63

02002993

 
avantor performance
N16808
Narrow Mouth Amber Glass Bottle 4L Each for $2,618.17
Description & Specifications

Specifications

Formula Weight 86.18
Chemical Name or Material Hexanes
Purity Grade ULTRA RESI-ANALYZED™
Assay 99% n-hexane
CAS 110-54-3
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.66kg
Flash Point −23.00°C
Quantity 1L
Molecular Formula CH3(CH2)4CH3

    For use in Gas Chromatography (GC); For Laboratory, Research, or Manufacturing Use