missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Hexanes, ≤97%, Extra Dry, mixture of isomers, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 610371000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexanes | |
Vapor Pressure: 160mbar at 20°C | |
C6H14 | |
MFCD00009520 | |
14, 4694 | |
86.18 | |
Extra Dry | |
100mL | |
AcroSeal™ Glass Bottle |
92112-69-1,110-54-3 | |
≤97% | |
CH3(CH2)4CH3 | |
01, 142 | |
Hex | |
≤97% | |
0.6590g/mL | |
Pass Test | |
50ppm max. |
RUO â Research Use Only