missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexanes, 98+%, extra pure, mixture of isomers, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 176660010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexanes | |
Mixture of Isomers | |
Colorless to Yellow | |
69.0°C | |
−22°C | |
Liquid or Solid | |
0.001 % max. | |
0.659 | |
CH3(CH2)4CH3 | |
0.31 mPa.s (20°C) | |
01, 142 | |
Hex | |
86.18 | |
Extra Pure |
-95.0°C | |
92112-69-1,110-54-3 | |
0.6590g/mL | |
1 L | |
Glass Bottle | |
1.3748 to 1.3810 | |
98+% | |
C6H14 | |
MFCD00009520 | |
0.05% Max. | |
14, 4694 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
≥98% |
RUO – Research Use Only