missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Hexane, mixture of isomers, Puriss., ≥85% isomers basis (GC), Honeywell

Puriss.,≥85% isomers basis (GC)

$103.51 - $103.51

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044905 View Documents Honeywell™
156131L
1L Glass Bottle Each for $103.51
 
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

110-54-3
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

110-54-3
MFCD02179311
1730733
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18g/mol
puriss.
0.075
Glass Bottle
3 (vs air)
Liquid
-154°C
Hexane, Mixture Of Isomers
C6H14
UN1208
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.178
CHEBI:29021
≥85% isomers basis (GC)
0.672g/mL at 25°C
437°F
n20/D 1.379(lit.)
−23°C (−9.4°F)
65°C
1L
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310-P370 + P378-P403 + P235

H225-H304-H315-H336-H361f-H373-H411

DOTInformation : class 3 - PG 2 - Hexanes