missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexane, CHROMASOLV™, ≥97%, LC-MS, Honeywell Riedel-de Haën™

CHROMASOLV™ LC-MS

Supplier:  Honeywell Chemicals 349861L

 View more versions of this product

Catalog No. 6001053



Description

Description

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

 

Specifications

Specifications

Hexane
0.659g/mL at 25°C
0.077
453°F
CH3(CH2)4CH3
5.2 psi (37.7°C), 132 mmHg (20°C), 256 mmHg (37.7°C)
1730733
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18g/mol
LC-MS CHROMASOLV™
110-54-3
1 L
−26°C (−14.8°F)
C6H14
MFCD02179311
UN1208
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.18
CHEBI:29021
≥97%

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC
Safety and Handling

Safety and Handling

P201-P210-P273-P301 + P310-P308 + P313-P331

RTECSNumber : MN9275000

SDS
Documents

Documents

Promotions

Promotions