Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexanamide, 99%

$44.50 - $118.53

Chemical Identifiers

Chemical Name or Material Hexanamide
CAS 628-02-4
Molecular Formula C6H13NO
Linear Formula CH3(CH2)4CONH2
Formula Weight 115.18
MDL Number MFCD00008046
Synonym Caproamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2592903

 
alfa aesar™
H2592903
1g Each for $44.50

AAH2592906

 
alfa aesar™
H2592906
5g Each for $118.53
Description & Specifications

Specifications

Formula Weight 115.18
Chemical Name or Material Hexanamide
TSCA TSCA
Assay 99%
Molecular Formula C6H13NO
Linear Formula CH3(CH2)4CONH2
MDL Number MFCD00008046
CAS 628-02-4
Synonym Caproamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MN7875000
Melting Point 100° to 102°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.