missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanamide 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 411530100

 View more versions of this product

Catalog No. AC411530100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
115.18
White to off-white
Glass bottle
Hexanamide
98%
C6H13NO
MFCD00008046
Flakes
10g
1% max. (K.F.)
Authentic
98%
CH3(CH2)4CONH2
628-02-4

Chemical Identifiers

628-02-4
C6H13NO
115.18
Hexanamide, 98%
98%
CH3(CH2)4CONH2
MFCD00008046
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-024-8

RTECSNumber : MN7875000

TSCA : TSCA