Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexanal dimethyl acetal, 98%

Chemical Identifiers

Chemical Name or Material Hexanal dimethyl acetal
CAS 1599-47-9
Molecular Formula C8H18O2
Linear Formula CH3(CH2)4CH(OCH3)2
Formula Weight 146.23
MDL Number MFCD00036508
UN Number UN1993
Synonym 1,1-Dimethoxyhexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1882422

 
alfa aesar™
A1882422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1882414

 
alfa aesar™
A1882414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.23
Chemical Name or Material Hexanal dimethyl acetal
TSCA TSCA
Assay 98%
Molecular Formula C8H18O2
Linear Formula CH3(CH2)4CH(OCH3)2
MDL Number MFCD00036508
CAS 1599-47-9
UN Number UN1993
Synonym 1,1-Dimethoxyhexane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 54°C (130°F)
Boiling Point 163° to 164°C
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.