Learn More
Hexamethylphosphorous triamide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 162480500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexamethylphosphorous triamide | |
-44°C | |
0.8900g/mL | |
26°C | |
96% min. (GC) | |
C6H18N3P | |
[(CH3)2N]3P | |
50 mL | |
0.89 | |
Solubility in water: reacts | |
CN(C)P(N(C)C)N(C)C | |
163.21 | |
163.2 | |
Liquid |
1608-26-0 | |
Colorless to Yellow | |
150°C | |
Authentic | |
Glass bottle | |
1.4650 to 1.467 | |
MFCD00008301 | |
01,431; 02,210; 04,247; 06,279; 09,235; 10,199 | |
hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide | |
XVDBWWRIXBMVJV-UHFFFAOYSA-N | |
N-[bis(dimethylamino)phosphanyl]-N-methylmethanamine | |
15355 | |
97% |
Chemical Identifiers
1608-26-0 | |
163.21 | |
XVDBWWRIXBMVJV-UHFFFAOYSA-N | |
15355 | |
CN(C)P(N(C)C)N(C)C |
C6H18N3P | |
MFCD00008301 | |
hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide | |
N-[bis(dimethylamino)phosphanyl]-N-methylmethanamine |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 216-534-4
Recommended Storage : product may develop some turbidity or precipitate
RUO – Research Use Only