missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethylenetetramine, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A172130B

 View more versions of this product

Catalog No. AAA172130B



Description

Description

Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

Solubility
Soluble in water, ethanol, chloroform and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Avoid conditions like flames, sparks and moisture.
Specifications

Specifications

Hexamethylenetetramine
∼280°C (sublimation)
250°C (482°F)
MFCD00006895
UN1328
Hygroscopic
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
VKYKSIONXSXAKP-UHFFFAOYSA-N
140.19
CHEBI:6824
≥99%
100-97-0
1.33
C6H12N4
1000 g
2018
14,5966
Soluble in water,ethanol,chloroform and acetone.
C1N2CN3CN1CN(C2)C3
4101
140.19

Chemical Identifiers

100-97-0
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
4101
C1N2CN3CN1CN(C2)C3
C6H12N4
MFCD00006895
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
CHEBI:6824
Safety and Handling

Safety and Handling

GHS H Statement
H228-H317
Flammable solid.
May cause an allergic skin reaction.

P210-P240-P241-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c

H228-H317-H500

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: HEXAMETHYLENETETRAMINE

EINECSNumber : 202-905-8

RTECSNumber : MN4725000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only