missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethylenetetramine, 98.5%, contains an anticaking agent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 232890025

 View more versions of this product

Catalog No. AC232890025



Description

Description

Specifications

Specifications

Hexamethylenetetramine
250°C
Plastic bottle
1.33
0.5% max. (K.F.)
260.0°C
98.5%
98.5%
MFCD00006895
01,427; 02,208; 04,243; 09,234
1,3,5,7-tetraazaadamantane, aminoform, formamine, hexamethylenamine, hexamethylenetetramine, hexamine, methenamin, methenamine, urotropin, urotropine
VKYKSIONXSXAKP-UHFFFAOYSA-N
140.19
CHEBI:6824
Crystalline Powder
1.3300g/mL
Authentic
Particle size (> 0.2mm):> 50%
263.0 °C
White
2.5kg
100-97-0
C6H12N4
01, 583
15, 6038
Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C)
C1N2CN3CN1CN(C2)C3
4101
140.19
98.5%

Chemical Identifiers

100-97-0
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
4101
C1N2CN3CN1CN(C2)C3
C6H12N4
MFCD00006895
1,3,5,7-tetraazaadamantane, aminoform, formamine, hexamethylenamine, hexamethylenetetramine, hexamine, methenamin, methenamine, urotropin, urotropine
CHEBI:6824
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Flammable solid.

GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

EINECSNumber : 202-905-8

RTECSNumber : MN4725000

TSCA : TSCA

SDS Documents