missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 232890010

 View more versions of this product

Catalog No. AC232890010


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexamethylenetetramine
100-97-0
White
250°C
98.5%
C6H12N4
1 kg
01,427; 02,208; 04,243; 09,234
15, 6038
263.0 °C
VKYKSIONXSXAKP-UHFFFAOYSA-N
140.19
CHEBI:6824
98.5%
98.5%
260.0°C
1.3300g/mL
Authentic
Plastic bottle
MFCD00006895
01, 583
1.33
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C)
C1N2CN3CN1CN(C2)C3
4101
140.19
Crystalline Powder

Chemical Identifiers

100-97-0
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
4101
C1N2CN3CN1CN(C2)C3
C6H12N4
MFCD00006895
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
CHEBI:6824
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Flammable solid.

GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

EINECSNumber : 202-905-8

RTECSNumber : MN4725000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only