Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexamethylditin, 97%

Chemical Identifiers

Chemical Name or Material Hexamethylditin
CAS 661-69-8
Molecular Formula C6H18Sn2
Linear Formula (CH3)3SnSn(CH3)3
Formula Weight 327.59
MDL Number MFCD00008277
UN Number UN2788
Beilstein 3600327
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1763803

  SDS
alfa aesar™
A1763803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1763806

  SDS
alfa aesar™
A1763806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1763814

  SDS
alfa aesar™
A1763814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 327.59
Chemical Name or Material Hexamethylditin
Boiling Point 85°C (45mmHg)
Melting Point 23°C
Assay 97%
Molecular Formula C6H18Sn2
Linear Formula (CH3)3SnSn(CH3)3
MDL Number MFCD00008277
CAS 661-69-8
Quantity 1g
UN Number UN2788
Beilstein 3600327
Flash Point 61°C (142°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H227
Fatal if swallowed.
Fatal in contact with skin.
Combustible liquid.
Odor Characteristic
Health Hazard 3 GHS P Statement
P210-P301+P310-P361-P302+P350-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
RTECS Number WH8280000

Applications
It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents, heat and air. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P361-P302+P350-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H227
Fatal if swallowed.
Fatal in contact with skin.
Combustible liquid.

2° to 8°C