missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethyldisiloxane, NMR grade, 99.7%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1697014

 View more versions of this product

Catalog No. AAL1697014



Description

Description

Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
Specifications

Specifications

100°C to 101°C
-67°C
25 g
(CH3)3SiOSi(CH3)3
UN1993
−2°C (28°F)
Slightly miscible with water.
C[Si](C)(C)O[Si](C)(C)C
162.379
CHEBI:78002
99.7%
Characteristic
0.764
Hexamethyldisiloxane
107-46-0
C6H18OSi2
MFCD00008265
1736258
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
UQEAIHBTYFGYIE-UHFFFAOYSA-N
trimethyl(trimethylsilyloxy)silane
24764
162.38
NMR
1.377

Chemical Identifiers

107-46-0
162.379
UQEAIHBTYFGYIE-UHFFFAOYSA-N
24764
trimethyl(trimethylsilyloxy)silane
C6H18OSi2
MFCD00008265
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
CHEBI:78002
C[Si](C)(C)O[Si](C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 203-492-7

RTECSNumber : JM9237000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only