Please login to your online account to display your discounted pricing

Hexamethyldisiloxane, 98+%, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  194790100

 View more versions of this product

Catalog No. AC194790100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Hexamethyldisiloxane
Color Clear colorless
Quantity 10mL
CAS 107-46-0
Density 0.7600g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay ≥98.0% (GC)
Assay Percent Range 98+%
Molecular Formula C6H18OSi2
Refractive Index 1.3765 to 1.3785 (20°C, 589nm)
Linear Formula (CH3)3SiOSi(CH3)3
Formula Weight 162.38
Fieser 08,240; 09,55; 10,196
Synonym HMDSO

Specifications

Chemical Name or Material Hexamethyldisiloxane
Assay ≥98.0% (GC)
Assay Percent Range 98+%
Molecular Formula C6H18OSi2
Linear Formula (CH3)3SiOSi(CH3)3
Formula Weight 162.38
Synonym HMDSO