missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexamethylcyclotrisiloxane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 216465000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hexamethylcyclotrisiloxane | |
541-05-9 | |
35°C | |
98% | |
C6H18O3Si3 | |
500g | |
04,III,1884 | |
hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185 | |
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1 | |
222.46 | |
222.47 |
98% | |
134.0°C | |
Authentic | |
Glass Bottle | |
MFCD00005943 | |
1325 | |
09,4559 | |
HTDJPCNNEPUOOQ-UHFFFAOYSA-N | |
hexamethyl-1,3,5,2,4,6-trioxatrisilinane | |
10914 | |
98% |
Safety and Handling
GHS H Statement
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Danger
EINECSNumber : 208-765-4
TSCA : TSCA
RUO â Research Use Only