Learn More
Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2302814
Description
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Incompatible in strong oxidizing agents.
Specifications
134°C | |
64°C to 66°C | |
25 g | |
MFCD00005943 | |
1766829 | |
hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185 | |
HTDJPCNNEPUOOQ-UHFFFAOYSA-N | |
2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane | |
10914 | |
97% |
Hexamethylcyclotrisiloxane | |
541-05-9 | |
C6H18O3Si3 | |
UN1325 | |
35°C (95°F) | |
Slightly soluble in water. | |
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1 | |
222.46 | |
222.47 | |
Moisture sensitive |
Chemical Identifiers
541-05-9 | |
222.46 | |
HTDJPCNNEPUOOQ-UHFFFAOYSA-N | |
10914 | |
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1 |
C6H18O3Si3 | |
MFCD00005943 | |
hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185 | |
2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 208-765-4
RTECSNumber : GZ4705000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only