missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexamethylbenzene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120570250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexamethylbenzene | |
87-85-4 | |
White to Yellow | |
Authentic | |
Glass bottle | |
C6(CH3)6 | |
25 g | |
01,426; 01,427; 02,207; 05,323; 06,273; 07,167; 09,234; 13,141; 14,175 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol | |
CC1=C(C(=C(C(=C1C)C)C)C)C | |
162.27 | |
CHEBI:39001 | |
98+% |
99+% | |
165°C to 168°C | |
264°C | |
98% min. (GC) | |
C12H18 | |
MFCD00008523 | |
05, 450 | |
hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene; | |
YUWFEBAXEOLKSG-UHFFFAOYSA-N | |
1,2,3,4,5,6-hexamethylbenzene | |
6908 | |
162.27 | |
Crystalline Powder, Crystals or Needles |
Chemical Identifiers
87-85-4 | |
162.27 | |
YUWFEBAXEOLKSG-UHFFFAOYSA-N | |
6908 | |
1,2,3,4,5,6-hexamethylbenzene |
C12H18 | |
MFCD00008523 | |
hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene; | |
CHEBI:39001 | |
CC1=C(C(=C(C(=C1C)C)C)C)C |
Safety and Handling
EINECSNumber : 201-777-0
RTECSNumber : DA3200000
TSCA : TSCA
RUO – Research Use Only