missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexamethylacetone, 98%

Manufacturer:  Alfa Aesar™ H5346414

 View more versions of this product

Catalog No. AAH5346414



Description

Description

Applications
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Hexamethylacetone
34°C (93°F)
152°C to 153°C
815-24-7
MFCD00008845
1701122
UIQGEWJEWJMQSL-UHFFFAOYSA-N
2,2,4,4-tetramethylpentan-3-one
13152
98%
0.824
1.419
25g
C9H18O
UN1224
2,2,4,4-tetramethyl-3-pentanone
CC(C)(C)C(=O)C(C)(C)C
142.242
142.24

Chemical Identifiers

815-24-7
142.242
UIQGEWJEWJMQSL-UHFFFAOYSA-N
13152
CC(C)(C)C(=O)C(C)(C)C
C9H18O
MFCD00008845
2,2,4,4-tetramethyl-3-pentanone
2,2,4,4-tetramethylpentan-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 212-419-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents