missing translation for 'onlineSavingsMsg'
Learn More

Hexamethonium bromide, 98+%

Nicotinic acetyl choline receptor antagonist | CAS: 55-97-0 | C12H30Br2N2 | 362.194 g/mol

$139.10 - $407.78

Chemical Identifiers

CAS 55-97-0
Molecular Formula C12H30Br2N2
Molecular Weight (g/mol) 362.194
MDL Number MFCD00011787
InChI Key FAPSXSAPXXJTOU-UHFFFAOYSA-L
Synonym hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide
PubChem CID 5938
IUPAC Name trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
SMILES C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6046614
View Documents
Thermo Scientific Chemicals
J6046614
25 g
Each for $139.10
Only null left
 
AAJ6046622
View Documents
Thermo Scientific Chemicals
J6046622
100 g
Each for $407.78
Only null left
 
Description

Description

Nicotinic acetyl choline receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

55-97-0
362.194
FAPSXSAPXXJTOU-UHFFFAOYSA-L
5938
C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]
C12H30Br2N2
MFCD00011787
hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide
trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide

Specifications

55-97-0
White
(CH3)3N(Br)(CH2)6N(Br)(CH3)3
25 g
14,4688
FAPSXSAPXXJTOU-UHFFFAOYSA-L
trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
5938
≥98%
Hexamethonium bromide
282°C to 285°C (decomposition)
C12H30Br2N2
MFCD00011787
3715749
hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide
C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]
362.194
362.19
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-249-7

RTECSNumber : BQ8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only