missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexakis(bromomethyl)benzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2201606

 View more versions of this product

Catalog No. AAB2201606



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexakis(bromomethyl)benzene
∼292°C (decomposition)
MFCD00182539
UN3261
XJOUCILNLRXRTF-UHFFFAOYSA-N
1,2,3,4,5,6-hexakis(bromomethyl)benzene
76540
98%
3095-73-6
C12H12Br6
5 g
hexakis bromomethyl benzene, hexa bromomethyl benzene, 1,2,3,4,5,6-hexakis bromomethyl benzene, benzene, hexakis bromomethyl, acmc-1aijn, 10.14272/xjoucilnlrxrtf-uhfffaoysa-n, benzene,1,2,3,4,5,6-hexakis bromomethyl, doi:10.14272/xjoucilnlrxrtf-uhfffaoysa-n
C(C1=C(C(=C(C(=C1CBr)CBr)CBr)CBr)CBr)Br
635.652
635.65

Chemical Identifiers

3095-73-6
635.652
XJOUCILNLRXRTF-UHFFFAOYSA-N
76540
C(C1=C(C(=C(C(=C1CBr)CBr)CBr)CBr)CBr)Br
C12H12Br6
MFCD00182539
hexakis bromomethyl benzene, hexa bromomethyl benzene, 1,2,3,4,5,6-hexakis bromomethyl benzene, benzene, hexakis bromomethyl, acmc-1aijn, 10.14272/xjoucilnlrxrtf-uhfffaoysa-n, benzene,1,2,3,4,5,6-hexakis bromomethyl, doi:10.14272/xjoucilnlrxrtf-uhfffaoysa-n
1,2,3,4,5,6-hexakis(bromomethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 221-443-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only