missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene, mass spectrometry standard, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC449172500

 View more versions of this product

Catalog No. AC449172500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Hexakis(1H, 1H, ωH-perfluoroalkoxy)phosphazene
Glass bottle
1.8
105809-15-2
hexa 2 h? methyl-1,3,5,2??,4??,6??-triazatriphosphinine, ultramark 1621, ultramark1621
CP1(=NP(=NP(=N1)(C)C)(C)C)C
2120
2120
1.8000g/mL
1.3490 to 1.3570
250mg
C6H18N3P3
DEBZEVJNWCNATM-MZWXYZOWSA-N
2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
44119860

Chemical Identifiers

105809-15-2
2120
hexa 2 h? methyl-1,3,5,2??,4??,6??-triazatriphosphinine, ultramark 1621, ultramark1621
2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
C6H18N3P3
DEBZEVJNWCNATM-MZWXYZOWSA-N
44119860
CP1(=NP(=NP(=N1)(C)C)(C)C)C
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS Documents