missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexahydrophthalimide 98.0+%, TCI America™

$28.21 - $155.95

Chemical Identifiers

CAS 1444-94-6
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00035737
InChI Key WLDMPODMCFGWAA-UHFFFAOYSA-N
Synonym 1,2-Cyclohexanedicarboximide
PubChem CID 73559
IUPAC Name 3a,4,5,6,7,7a-hexahydroisoindole-1,3-dione
SMILES C1CCC2C(C1)C(=O)NC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H136325G
View Documents Promotion Details
TCI America
H136325G
25 g
Each for $28.21
 
H1363250G
View Documents Promotion Details
TCI America
H1363250G
250 g
Each for $155.95
 
Description

Description

Specifications

Chemical Identifiers

1444-94-6
153.181
WLDMPODMCFGWAA-UHFFFAOYSA-N
73559
C1CCC2C(C1)C(=O)NC2=O
C8H11NO2
MFCD00035737
1,2-Cyclohexanedicarboximide
3a,4,5,6,7,7a-hexahydroisoindole-1,3-dione

Specifications

1444-94-6
White
MFCD00035737
1,2-Cyclohexanedicarboximide
C1CCC2C(C1)C(=O)NC2=O
153.181
153.18
Crystalline Powder
137°C
C8H11NO2
25 g
WLDMPODMCFGWAA-UHFFFAOYSA-N
3a,4,5,6,7,7a-hexahydroisoindole-1,3-dione
73559
≥98.0% (GC,T)
Hexahydrophthalimide
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'