Learn More
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 165395000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexahydro-4-methylphthalic anhydride | |
19438-60-9 | |
1.1600g/mL | |
>110°C | |
97.5% min. (GC) | |
C9H12O3 | |
MFCD00005927 | |
1.16 | |
Solubility in water: reacts with water | |
CC1CCC2C(C1)C(=O)OC2=O | |
168.19 | |
65 mPa.s (20°C) | |
98% |
98% | |
-29°C | |
120°C (1.0 Mbar) | |
Authentic | |
Glass bottle | |
1.4764 to 1.4784 | |
500 g | |
hexahydro-4-methylphthalic anhydride, 4-methylhexahydrophthalic anhydride, 5-methylhexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, 1,3-isobenzofurandione, hexahydro-5-methyl, 1,3-isobenzofurandione, hexahydromethyl, 4-methylcyclohexane-1,2-dicarboxylic anhydride, 5-methylhexahydrophthalic anhydride, 5-methyl-3a,4,5,6,7,7a-hexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers | |
FKBMTBAXDISZGN-UHFFFAOYSA-N | |
5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione | |
86876 | |
168.19 | |
Liquid |
Chemical Identifiers
19438-60-9 | |
168.19 | |
FKBMTBAXDISZGN-UHFFFAOYSA-N | |
86876 | |
CC1CCC2C(C1)C(=O)OC2=O |
C9H12O3 | |
MFCD00005927 | |
hexahydro-4-methylphthalic anhydride, 4-methylhexahydrophthalic anhydride, 5-methylhexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, 1,3-isobenzofurandione, hexahydro-5-methyl, 1,3-isobenzofurandione, hexahydromethyl, 4-methylcyclohexane-1,2-dicarboxylic anhydride, 5-methylhexahydrophthalic anhydride, 5-methyl-3a,4,5,6,7,7a-hexahydroisobenzofuran-1,3-dione, 4-methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers | |
5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 243-072-0
TSCA : TSCA
RUO – Research Use Only