missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexahydro-1,3,5-triphenyl-1,3,5-triazine, 98%

$22.64 - $70.95

Chemical Identifiers

CAS 91-78-1
Molecular Formula C21H21N3
Formula Weight 315.42
MDL Number MFCD00043590
Beilstein 91510
Synonym Formanil trimer; Hexahydro-1,3,5-triphenyl-s-triazine
Chemical Name or Material Hexahydro-1,3,5-triphenyl-1,3,5-triazine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0962009 SDS Alfa Aesar™
L0962009
10g Each for $22.64
AAL0962018 SDS Alfa Aesar™
L0962018
50g Each for $70.95
Specifications Description & Specifications

Chemical Identifiers

91-78-1
315.42
91510
Hexahydro-1,3,5-triphenyl-1,3,5-triazine
C21H21N3
MFCD00043590
Formanil trimer; Hexahydro-1,3,5-triphenyl-s-triazine

Specifications

190° to 192°C
91-78-1
315.42
91510
Hexahydro-1,3,5-triphenyl-1,3,5-triazine
10g
C21H21N3
MFCD00043590
Formanil trimer; Hexahydro-1,3,5-triphenyl-s-triazine
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA