missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexahydro-1,3,5-triphenyl-1,3,5-triazine, 98%

Manufacturer:  Alfa Aesar™ L0962009

 View more versions of this product

Catalog No. AAL0962009



Specifications

Specifications

Hexahydro-1,3,5-triphenyl-1,3,5-triazine
10g
C21H21N3
91510
VASMRQAVWVVDPA-UHFFFAOYSA-N
1,3,5-triphenyl-1,3,5-triazinane
7065
98%
190°C to 192°C
91-78-1
MFCD00043590
hexahydro-1,3,5-triphenyl-1,3,5-triazine, 1,3,5-triazine, hexahydro-1,3,5-triphenyl, anhydroformaldehyde aniline, 1,3,5-triphenylhexahydro-1,3,5-triazine, 1,3,5-triphenylhexahydro-s-triazine, unii-x3tz97444k, s-triazine, hexahydro-1,3,5-triphenyl, 1,3,5-triphenyl-1,3,5-triazaperhydroine, anhydroformaldehyde aniline van, methyleneaniline trimer
C1N(CN(CN1C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
315.42
315.42

Chemical Identifiers

91-78-1
315.42
VASMRQAVWVVDPA-UHFFFAOYSA-N
7065
C1N(CN(CN1C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
C21H21N3
MFCD00043590
hexahydro-1,3,5-triphenyl-1,3,5-triazine, 1,3,5-triazine, hexahydro-1,3,5-triphenyl, anhydroformaldehyde aniline, 1,3,5-triphenylhexahydro-1,3,5-triazine, 1,3,5-triphenylhexahydro-s-triazine, unii-x3tz97444k, s-triazine, hexahydro-1,3,5-triphenyl, 1,3,5-triphenyl-1,3,5-triazaperhydroine, anhydroformaldehyde aniline van, methyleneaniline trimer
1,3,5-triphenyl-1,3,5-triazinane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents