Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexafluoropropene trimer, 97%

$41.55 - $41.55

Chemical Identifiers

Chemical Name or Material Hexafluoropropene trimer
CAS 6792-31-0
Molecular Formula C9F18
Linear Formula [F3CC(F)=CF2]3
Formula Weight 450.07
MDL Number MFCD00043821
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1651506

 
alfa aesar™
L1651506
5g Each for $41.55
Description & Specifications

Specifications

Formula Weight 450.07
Assay 97%
Molecular Formula C9F18
Linear Formula [F3CC(F)=CF2]3
MDL Number MFCD00043821
CAS 6792-31-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Chemical Name or Material Hexafluoropropene trimer
Boiling Point 110° to 115°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.