Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexafluoropropene trimer, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1651506

 View more versions of this product

Catalog No. AAL1651506

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 450.07
Molecular Formula C9F18
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00043821
CAS 6792-31-0
Linear Formula [F3CC(F)=CF2]3
Assay 97%
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 110° to 115°C
Chemical Name or Material Hexafluoropropene trimer

Specifications

Chemical Name or Material Hexafluoropropene trimer
CAS 6792-31-0
Molecular Formula C9F18
Linear Formula [F3CC(F)=CF2]3
Formula Weight 450.07
MDL Number MFCD00043821
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents