Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexafluoroglutaryl chloride, 97%

Chemical Identifiers

Chemical Name or Material Hexafluoroglutaryl chloride
CAS 678-77-3
Molecular Formula C5Cl2F6O2
Formula Weight 276.95
MDL Number MFCD00039309
UN Number UN3265
Beilstein 1914561
Synonym Perfluoroglutaryl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1420006

 
alfa aesar™
A1420006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1420003

 
alfa aesar™
A1420003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1420014

 
alfa aesar™
A1420014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 276.95
MDL Number MFCD00039309
Quantity 5g
Molecular Formula C5Cl2F6O2
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
CAS 678-77-3
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
TSCA TSCA
Odor Acrid
Synonym Perfluoroglutaryl chloride
Assay 97%
Boiling Point 110° to 112°C
UN Number UN3265
Beilstein 1914561
Chemical Name or Material Hexafluoroglutaryl chloride
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.