Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexafluoroglutaric anhydride, 97%

Chemical Identifiers

Chemical Name or Material Hexafluoroglutaric anhydride
CAS 376-68-1
Molecular Formula C5F6O3
Formula Weight 222.04
MDL Number MFCD00006680
UN Number UN3265
Beilstein 210699
Synonym Perfluoroglutaric anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0279803

  SDS
alfa aesar™
L0279803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0279806

  SDS
alfa aesar™
L0279806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0279814

  SDS
alfa aesar™
L0279814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 222.04
Chemical Name or Material Hexafluoroglutaric anhydride
MDL Number MFCD00006680
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Molecular Formula C5F6O3
Synonym Perfluoroglutaric anhydride
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN3265
CAS 376-68-1
Beilstein 210699
Assay 97%
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 1 Danger
Quantity 1g
Boiling Point 71° to 72°C
Odor Pungent
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.