Learn More
Hexafluoroglutaric anhydride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0279806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexafluoroglutaric anhydride | |
1.652 | |
None | |
C5F6O3 | |
MFCD00006680 | |
UN3265 | |
Moisture sensitive | |
IHYAGCYJVNHXCT-UHFFFAOYSA-N | |
3,3,4,4,5,5-hexafluorooxane-2,6-dione | |
78995 | |
97% |
376-68-1 | |
71°C to 72°C | |
Pungent | |
1.324 | |
5 g | |
210699 | |
hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione | |
C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F | |
222.042 | |
222.04 |
Chemical Identifiers
376-68-1 | |
222.042 | |
IHYAGCYJVNHXCT-UHFFFAOYSA-N | |
78995 | |
C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F |
C5F6O3 | |
MFCD00006680 | |
hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione | |
3,3,4,4,5,5-hexafluorooxane-2,6-dione |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-811-8
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only