Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexafluoroglutaramide, 98%

$76.05 - $338.64

Chemical Identifiers

Chemical Name or Material Hexafluoroglutaramide
CAS 507-68-6
Molecular Formula C5H4F6N2O2
Formula Weight 238.1
MDL Number MFCD00014804
Synonym Perfluoroglutaramide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1674303

 
alfa aesar™
A1674303
1g Each for $76.05

AAA1674306

 
alfa aesar™
A1674306
5g Each for $338.64
Description & Specifications

Specifications

Formula Weight 238.1
Chemical Name or Material Hexafluoroglutaramide
Assay 98%
Molecular Formula C5H4F6N2O2
MDL Number MFCD00014804
CAS 507-68-6
Synonym Perfluoroglutaramide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 213° to 214°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.