Please login to your online account to display your discounted pricing

Hexafluorobenzene, 99%, Alfa Aesar™

$54.78 - $184.72

Chemical Identifiers

Chemical Name or Material Hexafluorobenzene
CAS 392-56-3
Physical Form Liquid
Assay Percent Range 99%
Molecular Formula C6F6
Formula Weight 186.06
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1150009

 
alfa aesar
A1150009*
10g Each for $54.78

AAA1150018

 
alfa aesar
A1150018
50g Each for $184.72
Description & Specifications

Specifications

Formula Weight 186.06
Quantity 10g
Chemical Name or Material Hexafluorobenzene
Physical Form Liquid
Assay Percent Range 99%
Molecular Formula C6F6
CAS 392-56-3
Density 1.616

Applications
Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. Further, it is used as a reference molecule to investigate tissue oxygenation in vivo studies. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.