Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexafluoro-2-(p-tolyl)-2-propanol, 97%

$99.59 - $99.59

Chemical Identifiers

Chemical Name or Material Hexafluoro-2-(p-tolyl)-2-propanol
CAS 2010-61-9
Molecular Formula C10H8F6O
Formula Weight 258.17
MDL Number MFCD00015357
Beilstein 2127760
Synonym alpha,alpha-Bis(trifluoromethyl)-4-methylbenzyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1812306

 
alfa aesar™
A1812306
5g Each for $99.59
Description & Specifications

Specifications

Formula Weight 258.17
Chemical Name or Material Hexafluoro-2-(p-tolyl)-2-propanol
TSCA TSCA
Assay 97%
Molecular Formula C10H8F6O
MDL Number MFCD00015357
CAS 2010-61-9
Beilstein 2127760
Synonym alpha,alpha-Bis(trifluoromethyl)-4-methylbenzyl alcohol
Boiling Point 181° to 182°C
Quantity 5g