Learn More
Hexaethylphosphorus Triamide, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 423081000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hexaethylphosphorus triamide | |
0.8900g/mL | |
58°C | |
94% min. (GC) | |
C12H30N3P | |
((C2H5)2N)3P | |
100g | |
tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine | |
FDIOSTIIZGWENY-UHFFFAOYSA-N | |
75292 | |
95% |
2283-11-6 | |
111°C (6.0mmHg) | |
Authentic | |
Glass bottle | |
1.4720 to 1.475 | |
MFCD00009041 | |
0.89 | |
Solubility in water: insoluble | |
247.36 | |
247.36 |
Chemical Identifiers
2283-11-6 | |
247.36 | |
FDIOSTIIZGWENY-UHFFFAOYSA-N | |
75292 |
C12H30N3P | |
MFCD00009041 | |
tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Warning
EINECSNumber : 218-920-8
RUO â Research Use Only