missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecyltrimethylammonium Tetrafluoroborate, TCI America™

$210.84 - $727.23

Chemical Identifiers

CAS 73257-08-6
Molecular Formula C19H42BF4N
Molecular Weight (g/mol) 371.356
MDL Number MFCD00144896
InChI Key ZYVAQFUOBZOOLY-UHFFFAOYSA-N
Synonym Cetyltrimethylammonium Tetrafluoroborate
PubChem CID 16213810
IUPAC Name hexadecyl(trimethyl)azanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CCCCCCCCCCCCCCCC[N+](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07345G View Documents TCI America
H07345G
5g Each for $727.23
 
H07341G View Documents TCI America
H07341G
1g Each for $210.84
 
Specifications

Chemical Identifiers

73257-08-6
371.356
ZYVAQFUOBZOOLY-UHFFFAOYSA-N
16213810
[B-](F)(F)(F)F.CCCCCCCCCCCCCCCC[N+](C)(C)C
C19H42BF4N
MFCD00144896
Cetyltrimethylammonium Tetrafluoroborate
hexadecyl(trimethyl)azanium;tetrafluoroborate

Specifications

White
73257-08-6
MFCD00144896
Cetyltrimethylammonium Tetrafluoroborate
[B-](F)(F)(F)F.CCCCCCCCCCCCCCCC[N+](C)(C)C
371.356
371.36
≥98.0% (N)
5g
C19H42BF4N
1759
ZYVAQFUOBZOOLY-UHFFFAOYSA-N
hexadecyl(trimethyl)azanium;tetrafluoroborate
16213810
Crystal-Powder at 20°C
Hexadecyltrimethylammonium Tetrafluoroborate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0184&(9)-0795&(9)-1971

TSCA : No