Please login to your online account to display your discounted pricing

Hexadecyltrimethylammonium Hydroxide, 25%in Methanol, ACROS Organics™

$585.67 - $585.67

Chemical Identifiers

Chemical Name or Material Hexadecyltrimethylammonium hydroxide
Name Note 25%in Methanol
CAS 505-86-2
Molecular Formula C19H43NO
Linear Formula CH3(CH2)15N(OH)(CH3)3
Formula Weight 301.55
MDL Number MFCD00041921
Synonym Cetyltrimethylammonium hydroxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC411420500

  SDS
thermo fisher scientific
AC411420500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC411421000

  SDS
thermo fisher scientific
411421000
Glass bottle 100g Each for $585.67
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 301.55
Quantity 50g
Color Clear colorless to pale yellow
EINECS Number 208-022-4
TSCA TSCA
Concentration 23 to 27 w/w% (in Methanol)
Density 0.83g/mL
Water 3% max. (K.F.)
Name Note 25%in Methanol
Molecular Formula C19H43NO
Linear Formula CH3(CH2)15N(OH)(CH3)3
MDL Number MFCD00041921
CAS 505-86-2
Synonym Cetyltrimethylammonium hydroxide
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes damage to organs.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material Hexadecyltrimethylammonium hydroxide

Phase transfer catalyst

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes damage to organs.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.