missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecyltrimethylammonium Hydroxide (10% in Water), TCI America™

$66.16 - $454.97

Chemical Identifiers

CAS 505-86-2
Molecular Formula C19H43NO
Molecular Weight (g/mol) 301.559
MDL Number MFCD00041921
InChI Key WJLUBOLDZCQZEV-UHFFFAOYSA-M
Synonym 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide 1:1, cetrimide inn:ban:jan, cetyl-trimethylammonium hydroxide, cetyltrimethylammonium hydroxide, hexadecyltrimethylammonium hydroxide, n,n,n-trimethyl-1-hexadecanaminiuhydroxide, n,n,n-trimethylhexadecan-1-aminium hydroxide, n-hexadecyltrimethylammonium hydroxide, unii-8q7d6895lv
PubChem CID 68166
IUPAC Name hexadecyl(trimethyl)azanium;hydroxide
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)C.[OH-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H097425ML View Documents Tci America
H097425ML
25mL Each for $66.16
 
H0974500ML View Documents Tci America
H0974500ML
500mL Each for $454.97
 
Specifications

Chemical Identifiers

505-86-2
301.559
WJLUBOLDZCQZEV-UHFFFAOYSA-M
68166
CCCCCCCCCCCCCCCC[N+](C)(C)C.[OH-]
C19H43NO
MFCD00041921
1-hexadecanaminium, n,n,n-trimethyl-, hydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide 1:1, cetrimide inn:ban:jan, cetyl-trimethylammonium hydroxide, cetyltrimethylammonium hydroxide, hexadecyltrimethylammonium hydroxide, n,n,n-trimethyl-1-hexadecanaminiuhydroxide, n,n,n-trimethylhexadecan-1-aminium hydroxide, n-hexadecyltrimethylammonium hydroxide, unii-8q7d6895lv
hexadecyl(trimethyl)azanium;hydroxide

Specifications

Yellow
505-86-2
MFCD00041921
1-hexadecanaminium, n,n,n-trimethyl-, hydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide 1:1, cetrimide inn:ban:jan, cetyl-trimethylammonium hydroxide, cetyltrimethylammonium hydroxide, hexadecyltrimethylammonium hydroxide, n,n,n-trimethyl-1-hexadecanaminiuhydroxide, n,n,n-trimethylhexadecan-1-aminium hydroxide, n-hexadecyltrimethylammonium hydroxide, unii-8q7d6895lv
CCCCCCCCCCCCCCCC[N+](C)(C)C.[OH-]
301.559
301.56
Hexadecyltrimethylammonium Hydroxide (10% in Water)
25mL
C19H43NO
3267
WJLUBOLDZCQZEV-UHFFFAOYSA-M
hexadecyl(trimethyl)azanium;hydroxide
68166
Clear Liquid at 20°C
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1971&(2)-0184

TSCA : Yes