missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecyltrimethylammonium Hexafluorophosphate, TCI America™

$152.78 - $598.91

Chemical Identifiers

CAS 101079-29-2
Molecular Formula C19H42F6NP
Molecular Weight (g/mol) 429.516
MDL Number MFCD00144894
InChI Key XIHYIALOMDSESV-UHFFFAOYSA-N
Synonym Cetyltrimethylammonium Hexafluorophosphate
PubChem CID 23500182
IUPAC Name hexadecyl(trimethyl)azanium;hexafluorophosphate
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)C.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07331G View Documents TCI America
H07331G
1g Each for $152.78
 
H073325G View Documents TCI America
H073325G
25g Each for $598.91
 
Specifications

Chemical Identifiers

101079-29-2
429.516
XIHYIALOMDSESV-UHFFFAOYSA-N
23500182
CCCCCCCCCCCCCCCC[N+](C)(C)C.F[P-](F)(F)(F)(F)F
C19H42F6NP
MFCD00144894
Cetyltrimethylammonium Hexafluorophosphate
hexadecyl(trimethyl)azanium;hexafluorophosphate

Specifications

White
1g
C19H42F6NP
Cetyltrimethylammonium Hexafluorophosphate
CCCCCCCCCCCCCCCC[N+](C)(C)C.F[P-](F)(F)(F)(F)F
429.516
429.52
≥98.0% (N)
185°C
101079-29-2
MFCD00144894
XIHYIALOMDSESV-UHFFFAOYSA-N
hexadecyl(trimethyl)azanium;hexafluorophosphate
23500182
Crystal-Powder at 20°C
Hexadecyltrimethylammonium Hexafluorophosphate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0184&(9)-0795&(9)-1971

TSCA : No