missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecylcyclohexane, TCI America™

$262.78 - $858.63

Chemical Identifiers

CAS 6812-38-0
Molecular Formula C22H44
Molecular Weight (g/mol) 308.594
MDL Number MFCD04038421
InChI Key NRHBFNBZAZZTAU-UHFFFAOYSA-N
Synonym 1-Cyclohexylhexadecane
PubChem CID 23250
IUPAC Name hexadecylcyclohexane
SMILES CCCCCCCCCCCCCCCCC1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09665G View Documents TCI America
H09665G
5g Each for $262.78
 
H096625G View Documents TCI America
H096625G
25g Each for $858.63
 
Specifications

Chemical Identifiers

6812-38-0
308.594
NRHBFNBZAZZTAU-UHFFFAOYSA-N
23250
CCCCCCCCCCCCCCCCC1CCCCC1
C22H44
MFCD04038421
1-Cyclohexylhexadecane
hexadecylcyclohexane

Specifications

White-Yellow
6812-38-0
MFCD04038421
NRHBFNBZAZZTAU-UHFFFAOYSA-N
hexadecylcyclohexane
23250
Crystal-Lump at 20°C
Hexadecylcyclohexane
5g
C22H44
1-Cyclohexylhexadecane
CCCCCCCCCCCCCCCCC1CCCCC1
308.594
308.59
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No