missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecyl Lactate, TCI America™

$39.72 - $187.92

Chemical Identifiers

CAS 35274-05-6
Molecular Formula C19H38O3
Molecular Weight (g/mol) 314.51
InChI Key WUKXMJCZWYUIRZ-UHFFFAOYSA-N
Synonym Cetyl Lactate, Hexadecyl 2-Hydroxypropionate, Lactic Acid Hexadecyl Ester
PubChem CID 37098
IUPAC Name hexadecyl 2-hydroxypropanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)C(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H1159500G View Documents TCI America
H1159500G
500g Each for $187.92
 
H115925G View Documents TCI America
H115925G
25g Each for $39.72
 
Specifications

Chemical Identifiers

35274-05-6
314.51
Cetyl Lactate, Hexadecyl 2-Hydroxypropionate, Lactic Acid Hexadecyl Ester
hexadecyl 2-hydroxypropanoate
C19H38O3
WUKXMJCZWYUIRZ-UHFFFAOYSA-N
37098
CCCCCCCCCCCCCCCCOC(=O)C(C)O

Specifications

White
500g
C19H38O3
WUKXMJCZWYUIRZ-UHFFFAOYSA-N
hexadecyl 2-hydroxypropanoate
37098
Crystal-Powder at 20°C
Hexadecyl Lactate
42°C
35274-05-6
Cetyl Lactate, Hexadecyl 2-Hydroxypropionate, Lactic Acid Hexadecyl Ester
CCCCCCCCCCCCCCCCOC(=O)C(C)O
314.51
314.51
≥75.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0951

TSCA : Yes