missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecyl Acrylate (stabilized with MEHQ), TCI America™

$105.42 - $608.06

Chemical Identifiers

CAS 13402-02-3
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
InChI Key PZDUWXKXFAIFOR-UHFFFAOYSA-N
Synonym Acrylic Acid Hexadecyl Ester
PubChem CID 83410
IUPAC Name hexadecyl prop-2-enoate
SMILES CCCCCCCCCCCCCCCCOC(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H116825G View Documents TCI America
H116825G
25g Each for $105.42
 
H1168250G View Documents TCI America
H1168250G
250g Each for $608.06
 
Specifications

Chemical Identifiers

13402-02-3
296.495
Acrylic Acid Hexadecyl Ester
hexadecyl prop-2-enoate
C19H36O2
PZDUWXKXFAIFOR-UHFFFAOYSA-N
83410
CCCCCCCCCCCCCCCCOC(=O)C=C

Specifications

Yellow
13402-02-3
Acrylic Acid Hexadecyl Ester
CCCCCCCCCCCCCCCCOC(=O)C=C
296.495
296.50
≥90.0% (GC)
25g
C19H36O2
PZDUWXKXFAIFOR-UHFFFAOYSA-N
hexadecyl prop-2-enoate
83410
Clear Liquid at 20°C
Hexadecyl Acrylate (stabilized with MEHQ)
Safety and Handling

Safety and Handling

TSCA : Yes