missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecanophenone, TCI America™

$117.64 - $352.92

Chemical Identifiers

CAS 6697-12-7
Molecular Formula C22H36O
Molecular Weight (g/mol) 316.53
MDL Number MFCD00008997
InChI Key IIOLAWJMOGLOIB-UHFFFAOYSA-N
Synonym Palmitophenone, Pentadecyl Phenyl Ketone
PubChem CID 81194
IUPAC Name 1-phenylhexadecan-1-one
SMILES CCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H059025G View Documents TCI America
H059025G
25g Each for $352.92
 
H05905G View Documents TCI America
H05905G
5g Each for $117.64
 
Specifications

Chemical Identifiers

6697-12-7
316.53
IIOLAWJMOGLOIB-UHFFFAOYSA-N
81194
CCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
C22H36O
MFCD00008997
Palmitophenone, Pentadecyl Phenyl Ketone
1-phenylhexadecan-1-one

Specifications

248°C
59°C
6697-12-7
MFCD00008997
IIOLAWJMOGLOIB-UHFFFAOYSA-N
1-phenylhexadecan-1-one
81194
Crystal-Powder at 20°C
Hexadecanophenone
White-Yellow
25g
C22H36O
Palmitophenone, Pentadecyl Phenyl Ketone
CCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
316.53
316.53
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No