Please login to your online account to display your discounted pricing

Hexadecanedioic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hexadecanedioic acid
Name Note 97%
CAS 505-54-4
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C16H30O4
Linear Formula HO2C(CH2)14CO2H
Formula Weight 286.4
Synonym Thapsic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175910010

  SDS
thermo fisher scientific
AC175910010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Fine Crystalline Powder
Formula Weight 286.4
Quantity 1g
Color White
EINECS Number 208-013-5
TSCA TSCA
Beilstein 02, 733
Chemical Name or Material Hexadecanedioic acid
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C16H30O4
Packaging Glass bottle
Linear Formula HO2C(CH2)14CO2H
CAS 505-54-4
Synonym Thapsic acid