Please login to your online account to display your discounted pricing

Hexadecane , Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Hexadecane
Assay Percent Range 99%
Molecular Formula CH3(CH2)14CH3
Formula Weight 226.45
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MKH223570

 
avantor performance
H22357
Glass 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.45
Assay Percent Range 99%
Molecular Formula CH3(CH2)14CH3
Chemical Name or Material Hexadecane
Packaging Glass
CAS 544-76-3
Quantity 100g