missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecane , Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance H22357

 View more versions of this product

Catalog No. MKH223570

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Hexadecane
100g
99%
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CCCCCCCCCCCCCCCC
226.448
CHEBI:45296
Glass
544-76-3
C16H34
DCAYPVUWAIABOU-UHFFFAOYSA-N
hexadecane
11006
226.45

Chemical Identifiers

544-76-3
226.448
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CHEBI:45296
CCCCCCCCCCCCCCCC
C16H34
DCAYPVUWAIABOU-UHFFFAOYSA-N
11006
hexadecane
Documents