Please login to your online account to display your discounted pricing

Hexadecane, ASTM Grade, 99.0% min., EMD Millipore

$719.74 - $719.74

Chemical Identifiers

Chemical Name or Material Hexadecane
Name Note For Gas Chromatography
CAS 544-76-3
Physical Form Liquid
Assay Percent Range 99.0% min.
Molecular Formula C16H34
Linear Formula CH3(CH2)14CH3
Formula Weight 226.44
Synonym Cetane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MHX02003

 
emd millipore corporation
HX02003
250g Each for $719.74
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 226.44
Color Clear colorless
Quantity 250g
Density 0.77g/cm3 (20°C)
Linear Formula CH3(CH2)14CH3
CAS 544-76-3
Synonym Cetane
Recommended Storage Store without restrictions
Chemical Name or Material Hexadecane
Name Note For Gas Chromatography
Assay Percent Range 99.0% min.
Molecular Formula C16H34
Vapor Pressure < - 0.1 hPa (20°C)
Safety and Handling

Store without restrictions